accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OO

CAS RN 321309-42-6

IUPAC Name: 2-methyl-5-phenylfuran-3-carbaldehyde
Synonyms: 2-methyl-5-phenylfuran-3-carbaldehyde
Molecular Weight (g/mol): 186.21
Molecular Formula: C12H10O2
InChi Key: QRBXYIYZWZGINC-UHFFFAOYSA-N
SMILES: CC1=C(C=O)C=C(O1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results