accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NClHNSNH2FFF

CAS RN 321430-48-2

IUPAC Name: [3-chloro-5-(trifluoromethyl)pyridin-2-yl]thiourea
Synonyms: 3-chloro-5-(trifluoromethyl)pyridin-2-ylthiourea
Molecular Weight (g/mol): 255.64
Molecular Formula: C7H5ClF3N3S
InChi Key: UDCNEYAYOVFLLX-UHFFFAOYSA-N
SMILES: NC(=S)NC1=C(Cl)C=C(C=N1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results