accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFFFFFONN

CAS RN 32477-35-3

IUPAC Name: 2,2,3,3,4,4,4-heptafluoro-1-(1H-imidazol-1-yl)butan-1-one
Synonyms: 2,2,3,3,4,4,4-heptafluoro-1-(imidazol-1-yl)butan-1-one
Molecular Weight (g/mol): 264.1
Molecular Formula: C7H3F7N2O
InChi Key: MSYHGYDAVLDKCE-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(=O)N1C=CN=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)