accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3248-91-7

ClCH3H2NCNH2H3CNH2CH3

CAS RN 3248-91-7

IUPAC Name: tris(4-amino-3-methylphenyl)methylium chloride
Synonyms: new fuchsine basic violet 2
Molecular Weight (g/mol): 365.91
Molecular Formula: C22H24ClN3
InChi Key: ORDGVBRMUJCHEO-UHFFFAOYSA-M
SMILES: [Cl-].CC1=CC(=CC=C1N)[C+](C1=CC=C(N)C(C)=C1)C1=CC=C(N)C(C)=C1
Quantity 
  • (2)
  • (2)
Color 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results