accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3264-82-2

IUPAC Name: nickel(II) acetylacetonate
Synonyms: nickel(II) acetylacetonate
Molecular Weight (g/mol): 258.93
Molecular Formula: C10H16NiO4
InChi Key: SHWZFQPXYGHRKT-FDGPNNRMSA-N
SMILES: [Ni].C\C(O)=C\C(C)=O.C\C(O)=C\C(C)=O
Percent Purity 
  • (4)
  • (3)
Physical Form 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results