CAS RN 326496-51-9
CAS RN 326496-51-9
IUPAC Name:
{4-[(acetyloxy)methyl]phenyl}boronic acid
Synonyms:
4-[(acetyloxy)methyl]phenylboronic acid
Molecular Weight (g/mol):
193.99
Molecular Formula:
C9H11BO4
InChi Key:
FPQABEOUQSYDPY-UHFFFAOYSA-N
SMILES:
CC(=O)OCC1=CC=C(C=C1)B(O)O
1
–
1
of
1
results
1
4-Acetoxymethylbenzeneboronic acid, 97%, Thermo Scientific™
CAS: 326496-51-9 Molecular Formula: C9H11BO4 Molecular Weight (g/mol): 193.993 MDL Number: MFCD08235082 InChI Key: FPQABEOUQSYDPY-UHFFFAOYSA-N Synonym: 4-acetoxymethyl phenyl boronic acid,4-acetoxymethyl phenylboronic acid,4-acetoxymethyl benzeneboronic acid,4-acetyloxy methyl phenylboronic acid,4-acetoxymethylbenzeneboronic acid,4-acetoxymethyl phenyl boronicacid,4-acetyloxy methyl phenyl boronic acid,4-acetoxymethyl benzeneboronic acid 97 PubChem CID: 10583943 IUPAC Name: [4-(acetyloxymethyl)phenyl]boronic acid SMILES: B(C1=CC=C(C=C1)COC(=O)C)(O)O