missing translation for 'onlineSavingsMsg'
Learn More

4-Acetoxymethylbenzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15439737
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15439737

Brand: Thermo Scientific Alfa Aesar H52966.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 326496-51-9
Molecular Formula C9H11BO4
Molecular Weight (g/mol) 193.993
MDL Number MFCD08235082
InChI Key FPQABEOUQSYDPY-UHFFFAOYSA-N
Synonym 4-acetoxymethyl phenyl boronic acid, 4-acetoxymethyl phenylboronic acid, 4-acetoxymethyl benzeneboronic acid, 4-acetyloxy methyl phenylboronic acid, 4-acetoxymethylbenzeneboronic acid, 4-acetoxymethyl phenyl boronicacid, 4-acetyloxy methyl phenyl boronic acid, 4-acetoxymethyl benzeneboronic acid 97
PubChem CID 10583943
IUPAC Name [4-(acetyloxymethyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)COC(=O)C)(O)O

Specifications

Melting Point 81°C to 83°C
Quantity 250 mg
Formula Weight 193.99
Percent Purity 97%
Chemical Name or Material 4-Acetoxymethylbenzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.