accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOHOFF

CAS RN 329014-60-0

IUPAC Name: 2,3-difluoro-4-methoxybenzoic acid
Synonyms: 2,3-difluoro-4-methoxybenzoic acid
Molecular Weight (g/mol): 188.13
Molecular Formula: C8H6F2O3
InChi Key: OBSJPUVORWCLNA-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(O)=O)C(F)=C1F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results