accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3291-03-0

H3COOH3COCH3OHNNH2

CAS RN 3291-03-0

IUPAC Name: 3,4,5-trimethoxybenzohydrazide
Synonyms: 3,4,5-trimethoxybenzohydrazide
Molecular Weight (g/mol): 226.23
Molecular Formula: C10H14N2O4
InChi Key: KQXHMNUXNHQSOW-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1OC)C(=O)NN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results