missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzhydrazide, 98%

Product Code. 11470531
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470531 25 g 25g
11460531 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470531 Supplier Thermo Scientific Alfa Aesar Supplier No. B20055.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3291-03-0 | C10H14N2O4 | 226.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3291-03-0
Molecular Formula C10H14N2O4
Molecular Weight (g/mol) 226.23
MDL Number MFCD00014758
InChI Key KQXHMNUXNHQSOW-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxybenzhydrazide, 3,4,5-trimethoxybenzoic acid hydrazide, 3,4,5-trimethoxybenzoylhydrazine, 3,4,5-trimethoxybenzoic hydrazide, 1,2,3-trimethoxybenzene-5-carbohydrazide, 3,4,5trimeophco, acmc-20ang8, maybridge1_004292, #, benzhydrazide, 3,4,5-trimethoxy
PubChem CID 76793
IUPAC Name 3,4,5-trimethoxybenzohydrazide
SMILES COC1=CC(=CC(OC)=C1OC)C(=O)NN

Specifications

Melting Point 158°C to 160°C
Quantity 25 g
Beilstein 484531
Formula Weight 226.23
Percent Purity 98%
Chemical Name or Material 3,4,5-Trimethoxybenzhydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.