accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONHNOOOO(E/Z)

CAS RN 329691-12-5

IUPAC Name: N'-{[4-(benzyloxy)-3-methoxyphenyl]methylidene}-2H-1,3-benzodioxole-5-carbohydrazide
Synonyms: N'-{[4-(benzyloxy)-3-methoxyphenyl]methylidene}-2H-1,3-benzodioxole-5-carbohydrazide
Molecular Weight (g/mol): 404.42
Molecular Formula: C23H20N2O5
InChi Key: ULOKADSYVZOTTL-UHFFFAOYSA-N
SMILES: COC1=C(OCC2=CC=CC=C2)C=CC(C=NNC(=O)C2=CC3=C(OCO3)C=C2)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results