missing translation for 'onlineSavingsMsg'
Learn More

FPR A14, TRC

Product Code. 30405827
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30405827 100 mg
30393658 50 mg
30404217 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30405827 Supplier Toronto Research Chemicals Supplier No. TRCF756515100MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 329691-12-5
Molecular Formula C23H20N2O5
Molecular Weight (g/mol) 404.42
Synonym 1,3-Benzodioxole-5-carboxylic Acid, [[3-methoxy-4-(phenylmethoxy)phenyl]methylene]hydrazide (9CI), FPR A14
SMILES COc1cc(\C=N/NC(=O)c2ccc3OCOc3c2)ccc1OCc4ccccc4

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C23H20N2O5/c1-27-21-11-17(7-9-19(21)28-14-16-5-3-2-4-6-16)13-24-25-23(26)18-8-10-20-22(12-18)30-15-29-20/h2-13H,14-15H2,1H3,(H,25,26)/b24-13-
Formula Weight 404.1372
Chemical Name or Material 1,3-Benzodioxolane-5-carboxylic acid 4'-benzyloxy-3'-methoxybenzylidene Hydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.