accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOOOOOHOHO

CAS RN 3301-79-9

IUPAC Name: 3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-6-carboxylic acid
Synonyms: carboxyfluorescein
Molecular Weight (g/mol): 376.32
Molecular Formula: C21H12O7
InChi Key: BZTDTCNHAFUJOG-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2C(=O)OC3(C2=C1)C1=CC=C(O)C=C1OC1=CC(O)=CC=C31
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (3)
  • (6)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (2)