accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CON

CAS RN 33021-53-3

IUPAC Name: 1-(quinolin-3-yl)ethan-1-one
Synonyms: 3-acetylquinoline
Molecular Weight (g/mol): 171.2
Molecular Formula: C11H9NO
InChi Key: VMZYRGLKJCRGST-UHFFFAOYSA-N
SMILES: CC(=O)C1=CN=C2C=CC=CC2=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results