accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 331001-62-8

IUPAC Name: 2-[2-(4-acetylphenyl)hydrazin-1-ylidene]-2-(3,3-dimethyl-3,4-dihydroisoquinolin-1-yl)acetamide
Synonyms: 2-[2-(4-acetylphenyl)hydrazin-1-ylidene]-2-(3,3-dimethyl-4H-isoquinolin-1-yl)acetamide
Molecular Weight (g/mol): 362.43
Molecular Formula: C21H22N4O2
InChi Key: ALJIEVIJBAJISI-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(NN=C(C(N)=O)C2=NC(C)(C)CC3=CC=CC=C23)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results