accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3319-15-1

H3COCH3HO

CAS RN 3319-15-1

IUPAC Name: 1-(4-methoxyphenyl)ethan-1-ol
Synonyms: 1-(4-methoxyphenyl)ethanol
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: IUUULXXWNYKJSL-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C(C)O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results