accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHOClCl

CAS RN 3321-92-4

IUPAC Name: 1-(3,5-dichloro-2-hydroxyphenyl)ethan-1-one
Synonyms: 1-(3,5-dichloro-2-hydroxyphenyl)ethanone
Molecular Weight (g/mol): 205.03
Molecular Formula: C8H6Cl2O2
InChi Key: CJFYGRLJDKWMDI-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(Cl)=CC(Cl)=C1O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results