accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

KSN

CAS RN 333-20-0

IUPAC Name: potassium cyanosulfanide
Synonyms: potassium thiocyanate rhocya potassium sulfocyanate potassium rhodanate potassium rhodanide KSCN
Molecular Weight (g/mol): 97.18
Molecular Formula: CKNS
InChi Key: ZNNZYHKDIALBAK-UHFFFAOYSA-M
SMILES: [K+].[S-]C#N
Molecular Weight (g/mol) 
  • (31)
  • (15)
Quantity 
  • (2)
  • (1)
  • (4)
  • (1)
  • (6)
  • (6)
  • (1)
  • (7)
  • (1)
  • (1)
  • (1)
  • (11)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (8)
  • (5)
  • (5)
  • (9)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (13)
  • (3)
  • (5)
  • (4)
Grade 
  • (2)
  • (8)
  • (4)
  • (2)
  • (7)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (6)
  • (12)
  • (2)
  • (12)
  • (6)
  • (3)
  • (2)
  • (31)
  • (15)
  • (2)
  • (1)
  • (4)
  • (1)
  • (6)
  • (6)
  • (1)
  • (7)
  • (1)
  • (1)
  • (1)
  • (11)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (8)
  • (5)
  • (5)
  • (9)
  • (3)
  • (3)
  • (1)
  • (13)
  • (3)
  • (5)
  • (4)
  • (2)
  • (8)
  • (4)
  • (2)
  • (7)
  • (3)
  • (1)
  • (2)