accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH3CSOOHNOCH3

CAS RN 333430-43-6

IUPAC Name: N-{4-[ethyl(2-methylphenyl)sulfamoyl]phenyl}acetamide
Synonyms: N-{4-[ethyl(2-methylphenyl)sulfamoyl]phenyl}acetamide
Molecular Weight (g/mol): 332.42
Molecular Formula: C17H20N2O3S
InChi Key: NGMYZQIEURPXLI-UHFFFAOYSA-N
SMILES: CCN(C1=CC=CC=C1C)S(=O)(=O)C1=CC=C(NC(C)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results