CAS RN 3343-45-1
CAS RN 3343-45-1
IUPAC Name:
1-(4-bromophenyl)-2-hydroxyethan-1-one
Synonyms:
1-(4-bromophenyl)-2-hydroxyethanone
Molecular Weight (g/mol):
215.05
Molecular Formula:
C8H7BrO2
InChi Key:
FGROGLJVXNYNQC-UHFFFAOYSA-N
SMILES:
OCC(=O)C1=CC=C(Br)C=C1
1
–
1
of
1
results
1
1-(4-Bromophenyl)-2-hydroxyethan-1-one, Thermo Scientific™
CAS: 3343-45-1 Molecular Formula: C8H7BrO2 Molecular Weight (g/mol): 215.046 MDL Number: MFCD00174275 InChI Key: FGROGLJVXNYNQC-UHFFFAOYSA-N Synonym: 1-4-bromophenyl-2-hydroxyethanone,1-4-bromophenyl-2-hydroxyethan-1-one,ethanone,1-4-bromophenyl-2-hydroxy,1-4-bromophenyl-2-hydroxy-1-ethanone,1-4-bromophenyl-2-hydroxyethane-1-one,p-bromphenacylalkohol,zlchem 616,bromophenylhydroxyethanone,p-bromo-a-hydroxy acetophenone,4-bromobenzoyl methanol PubChem CID: 520633 IUPAC Name: 1-(4-bromophenyl)-2-hydroxyethanone SMILES: C1=CC(=CC=C1C(=O)CO)Br