accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 335-08-0

CH3HONFFF

CAS RN 335-08-0

IUPAC Name: 3,3,3-trifluoro-2-hydroxy-2-methylpropanenitrile
Synonyms: 3,3,3-trifluoro-2-hydroxy-2-methylpropanenitrile
Molecular Weight (g/mol): 139.08
Molecular Formula: C4H4F3NO
InChi Key: XDCMNDCKYSQKAX-UHFFFAOYNA-N
SMILES: CC(O)(C#N)C(F)(F)F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)