accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 33558-36-0

CH3SiH3CFFFFFF

CAS RN 33558-36-0

IUPAC Name: [3,5-bis(trifluoromethyl)phenyl]dimethylsilyl
Synonyms: [3,5-bis(trifluoromethyl)phenyl]dimethylsilyl
Molecular Weight (g/mol): 271.26
Molecular Formula: C10H9F6Si
InChi Key: HJQABLGRCWBCDT-UHFFFAOYSA-N
SMILES: C[Si](C)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results