missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)phenyldimethylsilane, 95%

Product Code. 11318626
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11318626 5 g 5g
11308626 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318626 Supplier Thermo Scientific Alfa Aesar Supplier No. L17673.06
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 33558-36-0 | C10H9F6Si | 271.257 g/mol

3,5-Bis(trifluoromethyl)phenyldimethylsilane is used as the Intermediates of organic light-emitting diode and as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Bis(trifluoromethyl)phenyldimethylsilane is used as the Intermediates of organic light-emitting diode and as a pharmaceutical intermediate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33558-36-0
Molecular Formula C10H9F6Si
Molecular Weight (g/mol) 271.257
MDL Number MFCD02093948
InChI Key HJQABLGRCWBCDT-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenyl dimethylsilane, 3,5-bis trifluoromethyl phenyl-dimethyl-silicon, 3,5-bis-trifluoromethyl phenyldimethylsilane, 3,5-bis trifluoromethyl phenyl-dimethylsilicon, 3,5-bis trifluoromethyl phenyldimethylsilane
PubChem CID 11219490
IUPAC Name [3,5-bis(trifluoromethyl)phenyl]-dimethylsilicon
SMILES C[Si](C)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Boiling Point 40°C (1.5mmHg)
Quantity 5 g
Beilstein 2859742
Formula Weight 272.27
Percent Purity 95%
Refractive Index 1.4125
Density 1.23
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenyldimethylsilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.