accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 335649-61-1

BNOOH3CH3CCH3OSiCH3H3CCH3CH3H3CHOHO

CAS RN 335649-61-1

IUPAC Name: {1-[(tert-butoxy)carbonyl]-5-[(tert-butyldimethylsilyl)oxy]-1H-indol-2-yl}boronic acid
Synonyms: 1-(tert-butoxycarbonyl)-5-[(tert-butyldimethylsilyl)oxy]indol-2-ylboronic acid
Molecular Weight (g/mol): 391.35
Molecular Formula: C19H30BNO5Si
InChi Key: NXKIUYOUKLIWLP-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C(=CC2=CC(O[Si](C)(C)C(C)(C)C)=CC=C12)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results