accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HO

CAS RN 3360-41-6

IUPAC Name: 4-phenylbutan-1-ol
Synonyms: 4-phenylbutanol
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: LDZLXQFDGRCELX-UHFFFAOYSA-N
SMILES: OCCCCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results