accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OSN

CAS RN 3364-80-5

IUPAC Name: 1,3-thiazole-4-carbaldehyde
Synonyms: 1,3-thiazole-4-carbaldehyde
Molecular Weight (g/mol): 113.13
Molecular Formula: C4H3NOS
InChi Key: WRFKSVINLIQRKF-UHFFFAOYSA-N
SMILES: O=CC1=CSC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results