accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClClClCl2H2H

CAS RN 33685-54-0

IUPAC Name: tetrachloro(²H₂)ethane
Synonyms: tetrachloro(²H₂)ethane
Molecular Weight (g/mol): 169.85
Molecular Formula: C2H2Cl4
InChi Key: QPFMBZIOSGYJDE-QDNHWIQGSA-N
SMILES: [2H]C(Cl)(Cl)C([2H])(Cl)Cl
Molecular Weight (g/mol) 
  • (10)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (1)
  • (10)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)