CAS RN 338748-96-2
IUPAC Name:
(5-iodopyridin-2-yl)thiourea
Synonyms:
5-iodopyridin-2-ylthiourea
Molecular Weight (g/mol):
279.1
Molecular Formula:
C6H6IN3S
InChi Key:
CWKXZMWFEPLTCA-UHFFFAOYSA-N
SMILES:
NC(=S)NC1=NC=C(I)C=C1
1
–
1
of
1
results
1
N-(5-Iodo-2-pyridyl)thiourea, 97%, Thermo Scientific™
CAS: 338748-96-2 Molecular Formula: C6H6IN3S Molecular Weight (g/mol): 279.099 MDL Number: MFCD01315199 InChI Key: CWKXZMWFEPLTCA-UHFFFAOYSA-N Synonym: n-5-iodo-2-pyridinyl thiourea,5-iodopyridin-2-yl thiourea,thiourea, n-5-iodo-2-pyridinyl,n-5-iodo-2-pyridyl thiourea,n-5-iodopyridin-2-yl thiourea PubChem CID: 1477100 IUPAC Name: (5-iodopyridin-2-yl)thiourea SMILES: C1=CC(=NC=C1I)NC(=S)N