missing translation for 'onlineSavingsMsg'
Learn More

N-(5-Iodo-2-pyridyl)thiourea, 97%, Thermo Scientific™

Product Code. 15476907
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15476907 250 mg 250mg
15456907 1 g 1g
15466907 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15476907 Supplier Thermo Scientific Alfa Aesar Supplier No. H34009.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 338748-96-2
Molecular Formula C6H6IN3S
Molecular Weight (g/mol) 279.099
MDL Number MFCD01315199
InChI Key CWKXZMWFEPLTCA-UHFFFAOYSA-N
Synonym n-5-iodo-2-pyridinyl thiourea, 5-iodopyridin-2-yl thiourea, thiourea, n-5-iodo-2-pyridinyl, n-5-iodo-2-pyridyl thiourea, n-5-iodopyridin-2-yl thiourea
PubChem CID 1477100
IUPAC Name (5-iodopyridin-2-yl)thiourea
SMILES C1=CC(=NC=C1I)NC(=S)N

Specifications

Melting Point 219°C to 225°C
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 279.1
Percent Purity 97%
Chemical Name or Material N-(5-Iodo-2-pyridyl)thiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.