accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OCH3OH3CO

CAS RN 33884-41-2

IUPAC Name: 3-(ethoxymethylidene)pentane-2,4-dione
Synonyms: 3-(ethoxymethylidene)pentane-2,4-dione
Molecular Weight (g/mol): 156.18
Molecular Formula: C8H12O3
InChi Key: ITTXGKOHFZJUEX-UHFFFAOYSA-N
SMILES: CCOC=C(C(C)=O)C(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results