accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrHN

CAS RN 3389-21-7

IUPAC Name: 3-(2-bromoethyl)-1H-indole
Synonyms: 3-(2-bromoethyl)-1H-indole
Molecular Weight (g/mol): 224.1
Molecular Formula: C10H10BrN
InChi Key: NTLAICDKHHQUGC-UHFFFAOYSA-N
SMILES: BrCCC1=CNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results