missing translation for 'onlineSavingsMsg'
Learn More

Indolyl-3-(ethyl-Beta-bromide), TRC

Product Code. 30396041
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396041 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396041 Supplier Toronto Research Chemicals Supplier No. TRCI6280005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(2-Bromoethyl)indole
CAS 3389-21-7
Molecular Formula C10 H10 Br N
Quantity 5 g
Synonym 3-(2-Bromoethyl)indole, Indoramin Hydrochloride Imp. A (BP)
InChI Formula InChI=1S/C10H10BrN/c11-6-5-8-7-12-10-4-2-1-3-9(8)10/h1-4,7,12H,5-6H2
SMILES BrCCc1c[nH]c2ccccc12
IUPAC Name 3-(2-bromoethyl)-1H-indole
Molecular Weight (g/mol) 224.1
Formula Weight 222.9997
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.