accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 34157-83-0

CH3OHO(R)CH3(S)H3C(R)(R)CH3(S)H3C(S)CH3HOO

CAS RN 34157-83-0

IUPAC Name: (2R,4aS,6aS,12bR,14aS,14bR)-10-hydroxy-2,4a,6a,9,12b,14a-hexamethyl-11-oxo-1,2,3,4,4a,5,6,6a,11,12b,13,14,14a,14b-tetradecahydropicene-2-carboxylic acid
Synonyms: triterpene tripterine celastrol
Molecular Weight (g/mol): 450.62
Molecular Formula: C29H38O4
InChi Key: KQJSQWZMSAGSHN-JJWQIEBTSA-N
SMILES: CC1=C(O)C(=O)C=C2C1=CC=C1[C@@]2(C)CC[C@@]2(C)[C@@H]3C[C@@](C)(CC[C@]3(C)CC[C@]12C)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results