missing translation for 'onlineSavingsMsg'
Learn More

Celastrol, TRC

Product Code. 30365069
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365069 5 mg 5mg
30359332 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30365069 Supplier Toronto Research Chemicals Supplier No. TRCC2495005MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34157-83-0
Molecular Formula C29 H38 O4
Molecular Weight (g/mol) 450.61
Synonym 24,25,26-Trinoroleana-1(10),3,5,7-tetraen-29-oic acid, 3-hydroxy-9,13-dimethyl-2-oxo-, (9β,13α,14β,20α)-, 24-Nor-D:A-friedooleana-1(10),3,5,7-tetraen-29-oic acid, 3-hydroxy-2-oxo- (7CI,8CI), D:A-Friedo-24-noroleana-1(10),3,5,7-tetraen-29-oic acid, 3-hydroxy-2-oxo-, (20α)-, (9β,13α,14β,20α)-3-Hydroxy-9,13-dimethyl-2-oxo-24,25,26-trinoroleana-1(10),3,5,7-tetraen-29-oic acid, 2-Picenecarboxylic acid, 1,2,3,4,4a,5,6,6a,11,12b,13,14,14a,14b-tetradecahydro-10-hydroxy-2,4a,6a,9,12b,14a-hexamethyl-11-oxo-, (2R,4aS,6aS,12bR,14aS,14bR)-, 2-Picenecarboxylic acid, 1,2,3,4,4a,5,6,6a,11,12b,13,14,14a,14b-tetradecahydro-10-hydroxy-2,4a,6a,9,12b,14a-hexamethyl-11-oxo-, [2R-(2α,4aβ,6aβ,12bβ,14aα,14bβ)]-, Celastrol, NSC 70931, Tripterin
IUPAC Name (2R,4aS,6aR,6aS,14aS,14bR)-10-hydroxy-2,4a,6a,6a,9,14a-hexamethyl-11-oxo-1,3,4,5,6,13,14,14b-octahydropicene-2-carboxylic acid
SMILES C[C@@]12[C@]([C@@]3([H])[C@@](CC1)(CC[C@](C)(C3)C(O)=O)C)(CC[C@@]4(C2=CC=C5C4=CC(C(O)=C5C)=O)C)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C29H38O4/c1-17-18-7-8-21-27(4,19(18)15-20(30)23(17)31)12-14-29(6)22-16-26(3,24(32)33)10-9-25(22,2)11-13-28(21,29)5/h7-8,15,22,31H,9-14,16H2,1-6H3,(H,32,33)/t22-,25-,26-,27+,28-,29+/m1/s1
Formula Weight 450.277
Chemical Name or Material Celastrol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.