accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OONH2

CAS RN 344329-76-6

IUPAC Name: oxane-4-carboxamide
Synonyms: oxane-4-carboxamide
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: DGOYLVBDCVINQZ-UHFFFAOYSA-N
SMILES: NC(=O)C1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results