accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OFBr

CAS RN 345-94-8

IUPAC Name: 2-bromo-1-(4-fluorophenyl)propan-1-one
Synonyms: 2-bromo-1-(4-fluorophenyl)propan-1-one
Molecular Weight (g/mol): 231.06
Molecular Formula: C9H8BrFO
InChi Key: QKHHCXOSPQAQQI-UHFFFAOYNA-N
SMILES: CC(Br)C(=O)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results