accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 34598-49-7

IUPAC Name: 5-bromo-2,3-dihydro-1H-inden-1-one
Synonyms: 5-bromo-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 211.06
Molecular Formula: C9H7BrO
InChi Key: KSONICAHAPRCMV-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C(=O)CCC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results