accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 346696-69-3

CH3NHSOOOCH3

CAS RN 346696-69-3

IUPAC Name: 4-methoxy-N-[(4-methylphenyl)methyl]benzene-1-sulfonamide
Synonyms: 4-methoxy-N-[(4-methylphenyl)methyl]benzenesulfonamide
Molecular Weight (g/mol): 291.37
Molecular Formula: C15H17NO3S
InChi Key: PAIJRRXAEGQQNV-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)S(=O)(=O)NCC1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results