accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3468-01-7

IUPAC Name: 3-methyl-4-oxo-2-phenyl-4H-chromene-8-carboxylic acid
Synonyms: 8-carboxy-3-methylflavone
Molecular Weight (g/mol): 280.28
Molecular Formula: C17H12O4
InChi Key: KMMBBZOSQNLLMN-UHFFFAOYSA-N
SMILES: CC1=C(OC2=C(C=CC=C2C1=O)C(O)=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)