accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHFO

CAS RN 347-54-6

IUPAC Name: 5-fluoro-2-hydroxybenzaldehyde
Synonyms: 5-fluoro-2-hydroxybenzaldehyde
Molecular Weight (g/mol): 140.11
Molecular Formula: C7H5FO2
InChi Key: FDUBQNUDZOGOFE-UHFFFAOYSA-N
SMILES: OC1=CC=C(F)C=C1C=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)