accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 347-93-3

FOCl

CAS RN 347-93-3

IUPAC Name: 3-chloro-1-(4-fluorophenyl)propan-1-one
Synonyms: 3-chloro-1-(4-fluorophenyl)propan-1-one
Molecular Weight (g/mol): 186.61
Molecular Formula: C9H8ClFO
InChi Key: AAHQPLJUSLMHHR-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)CCCl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results