accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaH3CCH3CH3BrOHCH3CH3OH3CBrSOOO(Z)

CAS RN 34722-90-2

IUPAC Name: sodium 2-{[(1Z)-3-bromo-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene][3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]methyl}benzene-1-sulfonate
Synonyms: sodium 2-[(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)[(1Z)-3-bromo-5-isopropyl-2-methyl-4-oxocyclohexa-2,5-dien-1-ylidene]methyl]benzenesulfonate
Molecular Weight (g/mol): 646.37
Molecular Formula: C27H27Br2NaO5S
InChi Key: NMKFVGALBGZKGW-FKWCIMQXSA-M
SMILES: [Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (3)
  • (3)
Grade 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)