accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 348-27-6

IUPAC Name: 2-fluoro-4-hydroxybenzaldehyde
Synonyms: 2-fluoro-4-hydroxybenzaldehyde
Molecular Weight (g/mol): 140.11
Molecular Formula: C7H5FO2
InChi Key: ONRPXRPUBXXCCM-UHFFFAOYSA-N
SMILES: OC1=CC(F)=C(C=O)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results