accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

O

CAS RN 3481-02-5

IUPAC Name: cyclopropyl(phenyl)methanone
Synonyms: cyclopropyl phenyl ketone
Molecular Weight (g/mol): 146.19
Molecular Formula: C10H10O
InChi Key: PJRHFTYXYCVOSJ-UHFFFAOYSA-N
SMILES: O=C(C1CC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results