accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 348133-74-4

H3CNNHN

CAS RN 348133-74-4

IUPAC Name: 3-methyl-2-(piperazin-1-yl)quinoline
Synonyms: 3-methyl-2-(piperazin-1-yl)quinoline
Molecular Weight (g/mol): 227.31
Molecular Formula: C14H17N3
InChi Key: YUKANULGPVFVJO-UHFFFAOYSA-N
SMILES: CC1=C(N=C2C=CC=CC2=C1)N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results