CAS RN 34994-50-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Resorufin Sodium Salt ~90%, TRC
CAS: 34994-50-8 Molecular Formula: C12 H6 N O3 . Na Molecular Weight (g/mol): 235.17 IUPAC Name: sodium;7-oxophenoxazin-3-olate SMILES: [Na+].[O-]c1ccc2N=C3C=CC(=O)C=C3Oc2c1