Learn More
Resorufin Sodium Salt ~90%, TRC
Product Code. 30424271
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424271 1 g 1g
30409401 500 mg -
30409499 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30424271 Supplier Toronto Research Chemicals Supplier No. TRCR1465001G
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 34994-50-8
Molecular Formula C12 H6 N O3 . Na
Molecular Weight (g/mol) 235.17
IUPAC Name sodium;7-oxophenoxazin-3-olate
SMILES [Na+].[O-]c1ccc2N=C3C=CC(=O)C=C3Oc2c1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C12H7NO3.Na/c14-7-1-3-9-11(5-7)16-12-6-8(15)2-4-10(12)13-9;/h1-6,14H;/q;+1/p-1
Formula Weight 235.0245
Chemical Name or Material Resorufin Sodium Salt