accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3562-73-0

H3COH

CAS RN 3562-73-0

IUPAC Name: 1-{[1,1'-biphenyl]-4-yl}ethan-1-ol
Synonyms: 1-{[1,1'-biphenyl]-4-yl}ethanol
Molecular Weight (g/mol): 198.27
Molecular Formula: C14H14O
InChi Key: GOISDOCZKZYADO-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=C(C=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results