accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 359-13-7

HOFF

CAS RN 359-13-7

IUPAC Name: 2,2-difluoroethan-1-ol
Synonyms: 2,2-difluoroethanol
Molecular Weight (g/mol): 82.05
Molecular Formula: C2H4F2O
InChi Key: VOGSDFLJZPNWHY-UHFFFAOYSA-N
SMILES: OCC(F)F
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)