missing translation for 'onlineSavingsMsg'
Learn More

2,2-Difluoroethanol, 97%

Product Code. 11415826
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415826 1 g 1g
11425826 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415826 Supplier Thermo Scientific Alfa Aesar Supplier No. B22201.03
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 359-13-7 | C2H4F2O | 82.05 g/mol

2,2-Difluoroethanol is used as a chemical intermediates. It has wide applications in medicine and fluoropolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Difluoroethanol is used as a chemical intermediates. It has wide applications in medicine and fluoropolymers.

Solubility
soluble in chloroform, ethanol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 359-13-7
Molecular Formula C2H4F2O
Molecular Weight (g/mol) 82.05
MDL Number MFCD00042248
InChI Key VOGSDFLJZPNWHY-UHFFFAOYSA-N
Synonym 2,2-difluoroethan-1-ol, 2,2-difluoro-ethanol, ethanol, 2,2-difluoro, chf2ch2oh, 2,2-difluroethanol, 2,2-difluorethanol, 2,2-difluoro ethanol, 2,2-difluoroethylalcohol, difluoroethanol related, acmc-209ik0
PubChem CID 136187
IUPAC Name 2,2-difluoroethanol
SMILES C(C(F)F)O

Specifications

Melting Point -28°C
Density 1.296
Boiling Point 95°C to 96°C
Flash Point 3°C (37°F)
Refractive Index 1.3335
Quantity 1 g
UN Number UN1987
Beilstein 1731617
Solubility Information soluble in chloroform,ethanol.
Formula Weight 82.05
Percent Purity 97%
Chemical Name or Material 2,2-Difluoroethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.